
DL-3-Pyrrolidinol ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:40499-83-0
:
Formula:C₄H₉NO
MDL Number:MFCD00005256
:87.12 g/mol
Boiling Pt:179°C
EINECS:254-944-5
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together