Order Entry
Italy
ContactUsLinkComponent
(Perfluorohexyl)phenyliodonium trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
 
(Perfluorohexyl)phenyliodonium trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
  TCIAP1080-1G
 :  TCI
 :  
 
(Perfluorohexyl)phenyliodonium trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
  TCIAP1080-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (Potassium iodate Method):
    min. 98.0 %
  • Melting point:
    120.0 to 126.0 deg-C

 

 

 :
 :672.12 g/mol
 :GHS07
 :Refrigerator
 :77758-84-0
 :C₁₃H₅F₁₆IO₃S
 :MFCD00135119