0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
trans-2-Hexen-1-yl phenylacetate
trans-2-Hexen-1-yl phenylacetate
trans-2-Hexen-1-yl phenylacetate
TCIAP1045-25ML
:
TCI
:
trans-2-Hexen-1-yl phenylacetate
TCIAP1045-25ML
:
TCI
:
Pk:
25 mL
Seq:
2315
Purity (GC):
min. 85.0 %
:
:
218.30 g/mol
:
268-705-8
:
68133-78-8
:
C₁₄H₁₈O₂
:
MFCD00036543