Order Entry
Italy
ContactUsLinkComponent
Perfluorokerosene, super-high boiling for mass spectrometry
Perfluorokerosene, super-high boiling for mass spectrometry
Catalog # TCIAP1034-5G
Supplier:  TCI
CAS Number:  
undefined
Perfluorokerosene, super-high boiling for mass spectrometry
Catalog # TCIAP1034-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid to slightly cloudy liquid
  • Suitability for Mass-analysis calibratio:
    to pass test

Specification Test Results

About this item

 :
MDL Number:MFCD00132835