Order Entry
Italy
ContactUsLinkComponent
 
 
2,2-Pentamethylene-1,3-dioxolan-4-one ≥98.0% (by GC)
 :  TCI
 
 
2,2-Pentamethylene-1,3-dioxolan-4-one ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White or Colorless to Light yellow powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %

 

Frequently Bought Together