


Poly(methyl vinyl ether) 50% in methanol
TCIAP1007-500G
: TCI
:
- Pk:500 g
- Seq:3645
- Appearance:Colorless to Yellow to Orange clear liquid
- Evaporation residue:50.0 to 62.0 %
- Viscosity:4500 to 6500 mPa-s
- Absorbance:max. 0.150 (500.0nm)
- Absorbance2:max. 0.030 (600.0nm)
:
:1230
:3,II
:GHS02
:GHS07
:GHS08
:9003-09-2
:C₃H₆O
:MFCD00161528