1-(Pentafluorophenyl)ethanol
5 g
Catalog # TCIAP0925-5G
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: (1-Hydroxyethyl)pentafluorobenzene
- CAS Number:830-50-2
- Formula:C₈H₅F₅O
- MDL Number:MFCD00004510
- Molecular Weight:212.12 g/mol
- EINECS:212-597-7
Documents
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Freezing point:32.0 to 34.0 deg-C
Specifications
- Pk:5 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



