Order Entry
Italy
ContactUsLinkComponent
DMP (Dimethyl pimelimidate dihydrochloride) ≥98.0% (by titrimetric analysis) cross-linking agent, for peptide research
Irritant
DMP (Dimethyl pimelimidate dihydrochloride) ≥98.0% (by titrimetric analysis) cross-linking agent, for peptide research
Supplier:  TCI
Certificates
Irritant
DMP (Dimethyl pimelimidate dihydrochloride) ≥98.0% (by titrimetric analysis) cross-linking agent, for peptide research
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together