



Propylmagnesium bromide ∼27% ∼2 mol/L in THF
Catalog # TCIAP0880-250G
Supplier: TCI
CAS Number:
Specifications
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Concentration (sec Butanol method):26.0 to 30.0 w/w%
Specification Test Results
About this item
:
:147.30 g/mol
EINECS:213-162-4
UN:3399
ADR:4.3,I
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:927-77-5
Formula:C₃H₇BrMg
MDL Number:MFCD00058928