
Polyethylene glycol monooleyl ether n=∼10
Supplier: TCI
Certificates
About this item
CAS Number:9004-98-2
:
Formula:HO(CH₂CH₂O)nC₁₈H₃₅
MDL Number:MFCD00074369
EINECS:500-016-2
:GHS07
Specification Test Results
- The average value of n:8.7 to 10.8
Specification Test Results
Frequently Bought Together