Order Entry
Italy
ContactUsLinkComponent
Perfluorokerosene, low boiling for mass spectrometry
Flammable
Perfluorokerosene, low boiling for mass spectrometry
Catalog # TCIAP0690-10G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Perfluorokerosene, low boiling for mass spectrometry
Catalog # TCIAP0690-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Suitability for Mass-analysis calibratio:
    to pass test

Specification Test Results

About this item

 :
Flash Pt:46°C
UN:1993
ADR:3,III
 :GHS02
MDL Number:MFCD00132835