

Polyethylene glycol monododecyl ether n=∼25
Supplier: TCI
Certificates
About this item
CAS Number:9002-92-0
:
Formula:HO(CH₂CH₂O)nC₁₂H₂₅
MDL Number:MFCD00080891
Merck Index:14,07558
EINECS:500-002-6
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Freezing point:35.0 to 40.0 deg-C
- The average value of n:23.0 to 27.0
Specification Test Results
Frequently Bought Together