Phosphoenolpyruvic acid tris(cyclohexylammonium)salt, hydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:35556-70-8
:
Formula:C₃H₅O₆P·3C₆H₁₄N·xH₂O
MDL Number:MFCD00004258
:465.57 g/mol (as anhydrous)
Storage Temperature:Refrigerator
EINECS:252-618-7
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on anh.substance)
- Purity (Neutralization titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in Methanol:almost transparency
- Water:max. 5.0 %
Specification Test Results
Frequently Bought Together