Order Entry
Italy
ContactUsLinkComponent
 
2-Phenylethyl phenylacetate ≥98.0% (by GC)
  TCIAP0126-25G
 :  TCI
 :  
2-Phenylethyl phenylacetate ≥98.0% (by GC)
  TCIAP0126-25G
 :  TCI
 :  

 

  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White or Colorless to Light yellow powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :240.30 g/mol
 :330°C
 :203-013-1
 :102-20-5
 :C₁₆H₁₆O₂
 :MFCD00022049