Order Entry
Italy
ContactUsLinkComponent
Quercetin hydrate ≥96.0% (by HPLC and UV method)
Toxic
Quercetin hydrate ≥96.0% (by HPLC and UV method)
Catalog # TCIAP0042-25G
Supplier:  TCI
CAS Number:  
Toxic
Quercetin hydrate ≥96.0% (by HPLC and UV method)
Catalog # TCIAP0042-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 96.0 area%
  • Purity (UV-vis method):
    min. 96.0 % (calcd.on anh.substance)
  • Water:
    max. 15.0 %

Specification Test Results

About this item

 :
 :302.24 g/mol (anhy)
Merck Index:14,08034
EINECS:204-187-1
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:849061-97-8
Formula:C₁₅H₁₀O₇·xH₂O
MDL Number:MFCD00006828