Order Entry
Italy
ContactUsLinkComponent
(2-Oxo-1,3-dioxolan-4-yl)methyl methacrylate ≥98.0% (by GC) stabilised
(2-Oxo-1,3-dioxolan-4-yl)methyl methacrylate ≥98.0% (by GC) stabilised
Catalog # TCIAO0553-1G
Supplier:  TCI
CAS Number:  
(2-Oxo-1,3-dioxolan-4-yl)methyl methacrylate ≥98.0% (by GC) stabilised
Catalog # TCIAO0553-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g/mol
  • Seq:
    9220

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :186.16 g/mol
Boiling Pt:155°C
 :Stabilised with 4-methoxyphenol (MEHQ)
Storage Temperature:Refrigerator
CAS Number:13818-44-5
Formula:C₈H₁₀O₅
MDL Number:MFCD28964174