2,2'-Oxybis(N,N-di-n-octylacetamide) ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:342794-43-8
:
Formula:C₃₆H₇₂N₂O₃
MDL Number:MFCD23135286
:580.98 g/mol
Specification Test Results
- Appearance:Colorless to Red to Green clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together