Order Entry
Italy
ContactUsLinkComponent
 
Olsalazine Sodium ≥98.0% (by HPLC)
 :  TCI
 
Olsalazine Sodium ≥98.0% (by HPLC)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 95.0 %

 

Frequently Bought Together