2-Oxa-7-azaspiro[3.5]nonane hemioxalate ≥97.0% (by titrimetric analysis)
Catalog # TCIAO0509-250MG
Supplier: TCI
CAS Number:
Specifications
- Pk:250 mg
- Seq:2590
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (Nonaqueous Titration):min. 97.0 % (calcd.on free amine, dried substance)
- Oxalic Acid:25.0 to 44.0 %
- Water:max. 3.0 %
Specification Test Results
About this item
:
:344.4 g/mol
CAS Number:1429056-28-9
:MFCD22422281
Formula:C₁₆H₂₈N₂O₆
MDL Number:MFCD11035888