
n-Octyl acrylate ≥98.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:2499-59-4
:
Formula:C₁₁H₂₀O₂
MDL Number:MFCD00048935
:184.28 g/mol
Boiling Pt:57°C
:Stabilised with 4-methoxyphenol (MEHQ)
EINECS:219-696-4
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together