
2,2'-(2,2,3,3,4,4,5,5-Octafluorohexane-1,6-diyl)bis(oxirane) ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:791-22-0
:
Formula:C₁₀H₁₀F₈O₂
MDL Number:MFCD01320774
Boiling Pt:112°C
Flash Pt:133°C
:314.18 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together