4-n-Octyl-4H-dithieno[3,2-b:2',3'-d]pyrrole ≥98.0% (by GC)
: TCI
:141029-75-6
:
:C₁₆H₂₁NS₂
:MFCD26793506
:291.47 g/mol
:Refrigerator
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:33.0 to 37.0 deg-C
Frequently Bought Together