Order Entry
Italy
ContactUsLinkComponent
9-(9-Heptadecanyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥97.0% (by HPLC, total nitrogen)
9-(9-Heptadecanyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥97.0% (by HPLC, total nitrogen)
Supplier:  TCI
Certificates
undefined
9-(9-Heptadecanyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥97.0% (by HPLC, total nitrogen)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Melting point:
    144.0 to 148.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together