

1-Octen-3-one ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4312-99-6
:
Formula:C₈H₁₄O
MDL Number:MFCD00036558
:126.20 g/mol
Boiling Pt:60°C
Flash Pt:35°C
Storage Temperature:Refrigerator
EINECS:224-327-5
UN:1224
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together