Order Entry
Italy
ContactUsLinkComponent
2-Octen-4-one ≥96.0% (by GC)
Flammable
2-Octen-4-one ≥96.0% (by GC)
Catalog # TCIAO0360-5ML
Supplier:  TCI
CAS Number:  
Flammable
2-Octen-4-one ≥96.0% (by GC)
Catalog # TCIAO0360-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 96.0 %

Specification Test Results

About this item

 :
 :126.20 g/mol
Boiling Pt:66°C
Flash Pt:58°C
Storage Temperature:Refrigerator
EINECS:225-071-7
UN:1224
ADR:3,III
 :GHS02
CAS Number:4643-27-0
Formula:C₈H₁₄O
MDL Number:MFCD00061023