
(S)-(-)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol ≥99.0% (by GC)
: TCI
:65355-00-2
:
:C₂₀H₂₂O₂
:MFCD02093485
:294.39 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:165.0 to 168.0 deg-C
- Specific rotation [a]20/D:-48.0 to -52.0 deg (C=1, CHCl3)
- Solubility in Chloroform:almost transparency
Frequently Bought Together