
(R)-(+)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:65355-14-8
:
Formula:C₂₀H₂₂O₂
MDL Number:MFCD02093485
:294.39 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:166.0 to 170.0 deg-C
- Specific rotation [a]20/D:+48.0 to +52.0 deg (C=1, CHCl3)
- Solubility in Chloroform:almost transparency
Specification Test Results
Frequently Bought Together