Order Entry
Italy
ContactUsLinkComponent
 
 
 
4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl ≥95.0% (by GC, titration analysis) free radical
 :  TCI
 
 
 
4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl ≥95.0% (by GC, titration analysis) free radical
 :  TCI

 

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Purity (Iodometric Titration):
    min. 95.0 %
  • Melting point:
    35.0 to 39.0 deg-C

 

Frequently Bought Together