Order Entry
Italy
ContactUsLinkComponent
N-Octadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-18, SB-18) ≥97.0% (by total nitrogen basis)
Irritant
N-Octadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-18, SB-18) ≥97.0% (by total nitrogen basis)
Supplier:  TCI
Certificates
Irritant
N-Octadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-18, SB-18) ≥97.0% (by total nitrogen basis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 97.0 %

Specification Test Results

Frequently Bought Together