



n-Octylmagnesium bromide ∼22% ∼1 mol/L in THF
Catalog # TCIAO0240-250G
Supplier: TCI
CAS Number:
Specifications
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Concentration (sec Butanol method):21.0 to 25.0%
Specification Test Results
About this item
:
ADR:3,II
:217.43 g/mol
UN:2924
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:17049-49-9
Formula:C₈H₁₇BrMg
MDL Number:MFCD00191566