
2,6-Octadiyne ≥97.0% (by GC)
Catalog # TCIAO0194-0.1ML
Supplier: TCI
CAS Number:
Specifications
- Pk:0,1 mL
- Seq:2010
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to lump to clear liquid
- Purity (GC):min. 97.0 %
- Freezing point:25.0 to 28.0 deg-C
Specification Test Results
About this item
:
:106.17 g/mol
:GHS08
CAS Number:764-73-8
Formula:C₈H₁₀
MDL Number:MFCD00041620