2,2',3,3',5,5',6,6'-Octafluorobiphenyl ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3883-86-1
:
Formula:C₁₂H₂F₈
MDL Number:MFCD00000312
:298.14 g/mol
EINECS:223-418-7
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:83.0 to 86.0 deg-C
Specification Test Results
Frequently Bought Together