
Nefopam hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:23327-57-3
:
Formula:C₁₇H₂₀ClNO
MDL Number:MFCD00012750
:289.803 g/mol
Merck Index:14,06440
Melting Pt:251°C
Storage Temperature:Refrigerator
EINECS:245-585-5
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together