Order Entry
Italy
ContactUsLinkComponent
Naphtho[1,2-c:5,6-c']bis([1,2,5]oxadiazole) ≥95.0% (by HPLC)
Naphtho[1,2-c:5,6-c']bis([1,2,5]oxadiazole) ≥95.0% (by HPLC)
Catalog # TCIAN1137-200MG
Supplier:  TCI
CAS Number:  
Naphtho[1,2-c:5,6-c']bis([1,2,5]oxadiazole) ≥95.0% (by HPLC)
Catalog # TCIAN1137-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (HPLC):
    min. 95.0 area%
  • Elemental analysis (Nitrogen):
    25.00 to 27.00 %

Specification Test Results

About this item

 :
 :212.17 g/mol
CAS Number:1394221-73-8
Formula:C₁₀H₄N₄O₂