Noopept ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:157115-85-0
:
Formula:C₁₇H₂₂N₂O₄
MDL Number:MFCD00941121
:318.37 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:94.0 to 98.0 deg-C
- Specific rotation [a]20/D:-110 to -122 deg (C=0.4, CHCL3)
Specification Test Results
Frequently Bought Together