Naphtho[1,2-c:5,6-c']bis([1,2,5]thiadiazole) ≥98.0% (by HPLC)
Catalog # TCIAN1105-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Light yellow to Brown to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):21.80 to 24.10 %
- Melting point:246.0 to 250.0 deg-C
Specification Test Results
About this item
:
:244.29 g/mol
CAS Number:133546-47-1
Formula:C₁₀H₄N₄S₂
MDL Number:MFCD28579827