Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Neu5Ac alpha(2-6) N-Glycan
Neu5Ac alpha(2-6) N-Glycan ≥95.0% (by HPLC)
Neu5Ac alpha(2-6) N-Glycan ≥95.0% (by HPLC)
TCIAN1065-1MG
:
TCI
:
Neu5Ac alpha(2-6) N-Glycan ≥95.0% (by HPLC)
TCIAN1065-1MG
:
TCI
:
Pk:
1 mg
Seq:
2070
Appearance:
White to Almost white powder to crystal
Purity (HPLC):
min. 95.0 area%
:
:
2224.01 g/mol
:
Freezer
:
1125602-44-9
:
C₈₄H₁₃₈N₆O₆₂