NEO-823 ≥98.0% (by HPLC)
Catalog # TCIAN1056-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Nitrogen):7.00 to 8.00 %
Specification Test Results
About this item
:
:744.88 g/mol
CAS Number:1267603-73-5
Formula:C₄₄H₃₉F₃N₄O₂S