Nelfinavir mesylate hydrate ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:159989-65-8
:159989-64-7
Formula:C₃₂H₄₅N₃O₄S·CH₄O₃S·xH₂O
MDL Number:MFCD00931436
:663.89 g/mol (anhy)
Merck Index:14,06443
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Specific rotation [a]20/D:-109 to -113 deg (C=1, MeOH) (calcd.on anh.substance)
- Water:2.0 to 8.0 %
Specification Test Results
Frequently Bought Together