Order Entry
Italy
ContactUsLinkComponent
Neu5Ac alpha(2-6)Gal beta(1-4)GlcNAc-beta-ethylamine ≥98.0% (by HPLC)
Neu5Ac alpha(2-6)Gal beta(1-4)GlcNAc-beta-ethylamine ≥98.0% (by HPLC)
Catalog # TCIAN0950-5MG
Supplier:  TCI
CAS Number:  
undefined
Neu5Ac alpha(2-6)Gal beta(1-4)GlcNAc-beta-ethylamine ≥98.0% (by HPLC)
Catalog # TCIAN0950-5MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC) (CAD):
    min. 98.0 area%
  • Elemental analysis (Nitrogen):
    5.00 to 6.40 %

Specification Test Results

About this item

 :
 :739.66 g/mol
Storage Temperature:Freezer
Formula:C₂₇H₄₆N₃NaO₁₉