Order Entry
Italy
ContactUsLinkComponent
10-(2-Naphthyl)anthracene-9-boronic acid ≥98.0% (by HPLC) (contains varying amounts of Anhydride)
Irritant
10-(2-Naphthyl)anthracene-9-boronic acid ≥98.0% (by HPLC) (contains varying amounts of Anhydride)
Catalog # TCIAN0929-1G
Supplier:  TCI
CAS Number:  
undefined
Irritant
10-(2-Naphthyl)anthracene-9-boronic acid ≥98.0% (by HPLC) (contains varying amounts of Anhydride)
Catalog # TCIAN0929-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%

Specification Test Results

About this item

 :
 :348.21 g/mol
 :GHS07
CAS Number:597554-03-5
Formula:C₂₄H₁₇BO₂
MDL Number:MFCD11973625