
5-Norbornene-2-methylamine (mixture of isomers) ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:95-10-3
:
Formula:C₈H₁₃N
MDL Number:MFCD07369043
:123.20 g/mol
UN:2735
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 % (total of isomers)
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together