Naphthol AS-BI-β-D-Glucuronide ≥98.0% (by HPLC, total nitrogen) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:37-87-6
:
Formula:C₂₄H₂₂BrNO₉
MDL Number:MFCD00067162
:548.34 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- beta-Glucuronidase detection test:to pass test
Specification Test Results
Frequently Bought Together