4-Nitrophenyl-N-acetyl-b-D-galactosaminide
Supplier: TCI
Certificates
About this item
Synonyms: Gb5-β-pNP, 4-Nitrophenyl 2-Acetamido-2-deoxy-beta-D-galactopyranoside, 4-Nitrophenyl N-Acetyl-beta-D-galactosaminide, N-((2S,3R,4R,5R,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)tetrahydro-2H-pyran-3-yl)acetamide
CAS Number:14948-96-0
:
Formula:C₁₄H₁₈N₂O₈
MDL Number:MFCD00067360
Molecular Weight:342.31 g/mol
Melting Pt:207°C
Storage Temperature:Freezer
EINECS:239-024-3
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 95.0 %
- Specific rotation [a]20/D:+10.0 to +13.0 deg (C=1, DMSO)
Specification Test Results
Frequently Bought Together