

Nickel(II) trifluoromethanesulphonate ≥96.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:60871-84-3
:
Formula:C₂F₆NiO₆S₂
MDL Number:MFCD00673740
ADR:8,III
:356.82 g/mol
UN:1759
:GHS05
:GHS08
Specification Test Results
- Appearance:Light yellow to Yellow to Green powder to crystal
- Purity (Chelometric Titration):min. 96.0 %
Specification Test Results
Frequently Bought Together