Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Neu5Ac alpha(2-6)Gal beta(1-4)GlcNAc-beta-pNP
Neu5Ac-ɑ(2-6)Galβ(1-4)GlcNAc-β-pNP ≥93.0% (by HPLC)
Neu5Ac-ɑ(2-6)Galβ(1-4)GlcNAc-β-pNP ≥93.0% (by HPLC)
TCIAN0856-2MG
:
TCI
:
Neu5Ac-ɑ(2-6)Galβ(1-4)GlcNAc-β-pNP ≥93.0% (by HPLC)
TCIAN0856-2MG
:
TCI
:
Pk:
2 mg
Seq:
2145
Appearance:
White to Light yellow powder to crystal
Purity (HPLC):
min. 93.0 area%
:
:
795.70 g/mol
:
Freezer
:
501427-93-6
:
C₃₁H₄₅N₃O₂₁