Order Entry
Italy
ContactUsLinkComponent
Neu5Ac[1Me,4789Ac]α(2-3)Gal[246Bz]-β-MP ≥95.0% (by HPLC)
Neu5Ac[1Me,4789Ac]α(2-3)Gal[246Bz]-β-MP ≥95.0% (by HPLC)
Supplier:  TCI
Certificates
Neu5Ac[1Me,4789Ac]α(2-3)Gal[246Bz]-β-MP ≥95.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 95.0 area%

Specification Test Results

Related Information

Frequently Bought Together