
2-Naphthyl trifluoromethanesulphonate ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3857-83-8
:
Formula:C₁₁H₇F₃O₃S
MDL Number:MFCD00192341
:276.23 g/mol
Boiling Pt:87°C
Melting Pt:32°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together