Order Entry
Italy
ContactUsLinkComponent
 
Nortropinone hydrochloride ≥98.0% (by titrimetric analysis)
 :  TCI
 
Nortropinone hydrochloride ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %

 

Frequently Bought Together