Neu5Gc-ɑ(2-6)Gal-β-MP glycoside ≥97.0% (by HPLC)
Catalog # TCIAN0794-5MG
Supplier: TCI
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
Specification Test Results
About this item
:
:593.54 g/mol
Storage Temperature:Freezer
CAS Number:1072896-38-8
Formula:C₂₄H₃₅NO₁₆